ENA371464205 Enamine

2-[(5-{[(4-chlorophenyl)methyl]sulfanyl}-1,3,4-thiadiazol-2-yl)sulfanyl]acetohydrazide

  • MDL number MFCD07364298
  • Cas Number : 893726-46-0
  • Molecular Weight : 346.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-({5-[(4-chlorobenzyl)sulfanyl]-1,3,4-thiadiazol-2-yl}sulfanyl)acetohydrazide
Inchi Code 1S/C11H11ClN4OS3/c12-8-3-1-7(2-4-8)5-18-10-15-16-11(20-10)19-6-9(17)14-13/h1-4H,5-6,13H2,(H,14,17)
Inchi Key QTHBVOGKSDNUFV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110754
Availability details for
Quantity
  • Availability
  • Add to Cart