ENA371464206 Enamine

2-{[4-(4-methoxyphenyl)-5-(4-methylphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364300
  • Cas Number : 893726-52-8
  • Molecular Weight : 369.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110755
Availability details for
Quantity
  • Availability
  • Add to Cart