ENA371464207 Enamine

2-{[4-(4-chlorophenyl)-5-(4-methoxyphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364301
  • Cas Number : 893726-54-0
  • Molecular Weight : 389.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110756
Availability details for
Quantity
  • Availability
  • Add to Cart