ENA371464214 Enamine

2-[(5-{[(2-chlorophenyl)methyl]sulfanyl}-1,3,4-thiadiazol-2-yl)sulfanyl]acetohydrazide

  • MDL number MFCD07364308
  • Cas Number : 893726-70-0
  • Molecular Weight : 346.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110763
Availability details for
Quantity
  • Availability
  • Add to Cart