ENA371464217 Enamine

2-[(5-{[(4-methylphenyl)methyl]sulfanyl}-1,3,4-thiadiazol-2-yl)sulfanyl]acetohydrazide

  • MDL number MFCD07364310
  • Cas Number : 893726-76-6
  • Molecular Weight : 326.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110766
Availability details for
Quantity
  • Availability
  • Add to Cart