ENA371464220 Enamine

2-{[5-(4-tert-butylphenyl)-4-ethyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364312
  • Cas Number : 893726-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110769

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110769
Availability details for
Quantity
  • Availability
  • Add to Cart