ENA371464231 Enamine

2-{4-[(4-chlorophenyl)methyl]piperazin-1-yl}acetohydrazide

  • MDL number MFCD07364324
  • Cas Number : 893727-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110780
Availability details for
Quantity
  • Availability
  • Add to Cart