ENA371464237 Enamine

4-amino-5-(3,4,5-trimethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02729432
  • Cas Number : 31821-78-0
  • Molecular Weight : 282.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-5-(3,4,5-trimethoxyphenyl)-4H-1,2,4-triazole-3-thiol
Inchi Code 1S/C11H14N4O3S/c1-16-7-4-6(5-8(17-2)9(7)18-3)10-13-14-11(19)15(10)12/h4-5H,12H2,1-3H3,(H,14,19)
Inchi Key NALLUPJPVVQUMH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110786
Availability details for
Quantity
  • Availability
  • Add to Cart