ENA371464240 Enamine

4-amino-5-(3-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364329
  • Cas Number : 893727-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110789
Availability details for
Quantity
  • Availability
  • Add to Cart