ENA371464241 Enamine

2-{[4-(4-chlorophenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364333
  • Cas Number : 893727-15-6
  • Molecular Weight : 360.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110790
Availability details for
Quantity
  • Availability
  • Add to Cart