ENA371464245 Enamine

2-{[4-(4-ethoxyphenyl)-5-(pyridin-3-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364341
  • Cas Number : 893727-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110794

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110794
Availability details for
Quantity
  • Availability
  • Add to Cart