ENA371464246 Enamine

4-amino-5-[3-(propan-2-yloxy)phenyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364343
  • Cas Number : 893727-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110795
Availability details for
Quantity
  • Availability
  • Add to Cart