ENA371464248 Enamine

2-{[4-(2-methylphenyl)-5-phenyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364344
  • Cas Number : 893727-47-4
  • Molecular Weight : 339.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{[4-(2-methylphenyl)-5-phenyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide
Inchi Code 1S/C17H17N5OS/c1-12-7-5-6-10-14(12)22-16(13-8-3-2-4-9-13)20-21-17(22)24-11-15(23)19-18/h2-10H,11,18H2,1H3,(H,19,23)
Inchi Key VAPTZSCZORLGDJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110797
Availability details for
Quantity
  • Availability
  • Add to Cart