ENA371464252 Enamine

ethyl 5-amino-1-(4-ethoxyphenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD07388501
  • Cas Number : 30829-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110801
Availability details for
Quantity
  • Availability
  • Add to Cart