ENA371464258 Enamine

1-(4-methylphenyl)-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD09959861
  • Cas Number : 54738-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110807
Availability details for
Quantity
  • Availability
  • Add to Cart