ENA371464263 Enamine

2-[2-(4-methoxyphenyl)-2-oxoethyl]-5,5-dimethylcyclohexane-1,3-dione

  • MDL number MFCD07364349
  • Cas Number : 112404-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110812
Availability details for
Quantity
  • Availability
  • Add to Cart