ENA371464265 Enamine

5,5-dimethyl-2-[2-(4-methylphenyl)-2-oxoethyl]cyclohexane-1,3-dione

  • MDL number MFCD07364350
  • Cas Number : 893727-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110814
Availability details for
Quantity
  • Availability
  • Add to Cart