ENA371464277 Enamine

ethyl 5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00973905
  • Cas Number : 14678-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110826
Availability details for
Quantity
  • Availability
  • Add to Cart