ENA371464285 Enamine

3-methyl-1-(2-methylphenyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00461405
  • Cas Number : 29211-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110834
Availability details for
Quantity
  • Availability
  • Add to Cart