ENA371464287 Enamine

4-amino-5-(3-phenyl-1H-pyrazol-5-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364356
  • Cas Number : 366007-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259-260
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110836
Availability details for
Quantity
  • Availability
  • Add to Cart