ENA371464288 Enamine

4-amino-5-[3-(4-ethylphenyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364357
  • Cas Number : 1028451-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110837
Availability details for
Quantity
  • Availability
  • Add to Cart