ENA371464294 Enamine

2-{[5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD01571789
  • Cas Number : 312522-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110843
Availability details for
Quantity
  • Availability
  • Add to Cart