ENA371464302 Enamine

3-(thiophen-2-yl)-1H-pyrazole-5-carbohydrazide

  • MDL number MFCD01913972
  • Cas Number : 92352-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110851
Availability details for
Quantity
  • Availability
  • Add to Cart