ENA371464305 Enamine

6-bromo-5-methyl-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD00184166
  • Cas Number : 28279-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110854

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110854
Availability details for
Quantity
  • Availability
  • Add to Cart