ENA371464315 Enamine

3-({1,3-dibenzyl-2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-5-yl}carbamoyl)propanoic acid

  • MDL number MFCD07364268
  • Cas Number : 893725-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110706
Availability details for
Quantity
  • Availability
  • Add to Cart