ENA371464318 Enamine

4-amino-5-[3-(4-methylphenyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364365
  • Cas Number : 366007-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110867
Availability details for
Quantity
  • Availability
  • Add to Cart