ENA371464320 Enamine

4-amino-5-[3-(4-chlorophenyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364367
  • Cas Number : 1028417-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110869
Availability details for
Quantity
  • Availability
  • Add to Cart