ENA371464335 Enamine

5-methyl-3-(pyridin-4-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD06589776
  • Cas Number : 90771-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110884
Availability details for
Quantity
  • Availability
  • Add to Cart