ENA371464336 Enamine

5-methyl-3-(pyridin-3-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD06589775
  • Cas Number : 842958-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110885
Availability details for
Quantity
  • Availability
  • Add to Cart