ENA371464337 Enamine

5-methyl-3-(pyridin-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD06589774
  • Cas Number : 842958-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110886
Availability details for
Quantity
  • Availability
  • Add to Cart