ENA371464347 Enamine

2-phenyl-N-(2-phenylethyl)acetamide

  • MDL number MFCD00043534
  • Cas Number : 5460-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-110897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110897
Availability details for
Quantity
  • Availability
  • Add to Cart