ENA371464351 Enamine

(2-fluoro-4-methylphenyl)hydrazine hydrochloride

  • MDL number MFCD06738785

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110901

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110901
Availability details for
Quantity
  • Availability
  • Add to Cart