ENA371464369 Enamine

2-(4-methylpiperazin-1-yl)acetamide

  • MDL number MFCD06081244
  • Cas Number : 83808-21-3
  • Molecular Weight : 157.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 124-126
Iupac Name 2-(4-methyl-1-piperazinyl)acetamide
Inchi Code 1S/C7H15N3O/c1-9-2-4-10(5-3-9)6-7(8)11/h2-6H2,1H3,(H2,8,11)
Inchi Key VRZZMRYGHFNKEN-UHFFFAOYSA-N

Description

Melting Point 124-126
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110919

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-110919
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart