ENA371464371 Enamine

2-[4-(4-chlorophenyl)piperazin-1-yl]acetonitrile

  • MDL number MFCD09959869
  • Cas Number : 91494-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110921
Availability details for
Quantity
  • Availability
  • Add to Cart