ENA371464372 Enamine

2-(pyridin-4-yl)-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD00171525
  • Cas Number : 10478-99-6
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110922
Availability details for
Quantity
  • Availability
  • Add to Cart