ENA371464374 Enamine

1,3-diethyl 2-{[(2,4-dimethylphenyl)amino]methylidene}propanedioate

  • MDL number MFCD00136259
  • Cas Number : 104007-06-9
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110924
Availability details for
Quantity
  • Availability
  • Add to Cart