ENA371464382 Enamine

4-methyl-3,5-dinitrobenzohydrazide

  • MDL number MFCD09959872
  • Cas Number : 209341-86-6
  • Molecular Weight : 240.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110932
Availability details for
Quantity
  • Availability
  • Add to Cart