ENA371464383 Enamine

5-[(2-chloro-6-fluorophenyl)methyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD00973901
  • Cas Number : 874607-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110933
Availability details for
Quantity
  • Availability
  • Add to Cart