ENA371464384 Enamine

3-(2,5-dimethoxyphenyl)-1H-pyrazole-5-carbohydrazide

  • MDL number MFCD02668327
  • Cas Number : 1037690-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110934

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110934
Availability details for
Quantity
  • Availability
  • Add to Cart