ENA371464385 Enamine

3-({1,3-dimethyl-2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-5-yl}carbamoyl)propanoic acid

  • MDL number MFCD09959855
  • Cas Number : 1110717-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110710
Availability details for
Quantity
  • Availability
  • Add to Cart