ENA371464387 Enamine

1-(morpholin-4-yl)-2-[4-(propan-2-yl)phenyl]ethane-1-thione

  • MDL number MFCD00230092
  • Cas Number : 14182-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110937
Availability details for
Quantity
  • Availability
  • Add to Cart