ENA371464408 Enamine

4-(cyclopentylamino)-1-methylpiperidine-4-carbonitrile

  • MDL number MFCD09959882
  • Cas Number : 1018435-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110960
Availability details for
Quantity
  • Availability
  • Add to Cart