ENA371464420 Enamine

4-(4-chlorobenzenesulfonyl)piperazine-1-carbaldehyde

  • MDL number MFCD01185485
  • Cas Number : 928937-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110972
Availability details for
Quantity
  • Availability
  • Add to Cart