ENA371464421 Enamine

4-(4-bromobenzenesulfonyl)piperazine-1-carbaldehyde

  • MDL number MFCD04331021
  • Cas Number : 928888-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110973
Availability details for
Quantity
  • Availability
  • Add to Cart