ENA371464423 Enamine

methyl 2-{[4-(4-methylphenyl)-3-oxo-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-5-yl]sulfanyl}acetate

  • MDL number MFCD01907317
  • Cas Number : 312625-68-6
  • Molecular Weight : 400.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl {[3-(4-methylphenyl)-4-oxo-3,4,5,6,7,8-hexahydro[1]benzothieno[2,3-d]pyrimidin-2-yl]thio}acetate
Inchi Code 1S/C20H20N2O3S2/c1-12-7-9-13(10-8-12)22-19(24)17-14-5-3-4-6-15(14)27-18(17)21-20(22)26-11-16(23)25-2/h7-10H,3-6,11H2,1-2H3
Inchi Key VWIPGSBRLLDKFI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110975
Availability details for
Quantity
  • Availability
  • Add to Cart