ENA371464424 Enamine

methyl 2-{[3-(4-methylphenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]sulfanyl}acetate

  • MDL number MFCD03417407
  • Cas Number : 28831-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110976

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110976
Availability details for
Quantity
  • Availability
  • Add to Cart