ENA371464425 Enamine

methyl 2-{[3-(4-chlorophenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]sulfanyl}acetate

  • MDL number MFCD05154684
  • Cas Number : 28831-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110977
Availability details for
Quantity
  • Availability
  • Add to Cart