ENA371464435 Enamine

3-(3,6-dichloro-9H-carbazol-9-yl)propanehydrazide

  • MDL number MFCD00572877
  • Cas Number : 300816-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110987

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110987
Availability details for
Quantity
  • Availability
  • Add to Cart