ENA371464442 Enamine

5,5-dimethyl-2-(3-oxo-1,3-diphenylpropyl)cyclohexane-1,3-dione

  • MDL number MFCD00520712
  • Cas Number : 53852-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110994
Availability details for
Quantity
  • Availability
  • Add to Cart