ENA371464456 Enamine

2,4-dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole

  • MDL number MFCD01763614
  • Cas Number : 439107-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111008

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111008
Availability details for
Quantity
  • Availability
  • Add to Cart